Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THAP6
Entrez Gene ID
152815
Gene Name
THAP domain containing 6
Gene Aliases
-
Chromosome Location
Chr.4: 75513951 - 75547334 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144721.5 NP_653322.1 3 - 4 393 104
XM_005262772.2 XP_005262829.1 2 - 3 288 104
XM_005262774.3 XP_005262831.1 3 - 4 395 104
XM_006714109.3 XP_006714172.1 3 - 4 395 104
XM_011531666.2 XP_011529968.1 3 - 4 830 104
XM_011531667.2 XP_011529969.1 3 - 4 353 104
XM_017007801.1 XP_016863290.1 3 - 4 395 104
GenBank mRNA AK303658.1 - 3 - 4 353 104
AK308914.1 - 2 - 3 253 104
AK310575.1 - 2 - 3 257 104
AK313765.1 - 3 - 4 777 104
AL833350.1 - 3 - 4 353 104
BC022989.2 - 2 - 3 307 104
DC308535.1 - 3 - 4 394 104

View Full Product Details