Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PACRGL
- Entrez Gene ID
-
133015
- Gene Name
- PARK2 coregulated like
- Gene Aliases
-
C4orf28
- Chromosome Location
-
Chr.4: 20696282 - 20757766 on Build GRCh38
View Full Product Details