The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282941.1 | NP_001269870.1 | 1 - 2 | 314 | 137 |
NM_145307.3 | NP_660350.2 | 1 - 2 | 314 | 137 | |
XM_011539456.2 | XP_011537758.1 | 1 - 2 | 330 | 137 | |
XM_011539457.2 | XP_011537759.1 | 1 - 2 | 330 | 137 | |
XM_011539460.2 | XP_011537762.1 | 1 - 2 | 330 | 137 | |
XM_017015842.1 | XP_016871331.1 | 1 - 2 | 291 | 137 | |
XM_017015843.1 | XP_016871332.1 | 1 - 2 | 330 | 137 | |
XM_017015844.1 | XP_016871333.1 | 1 - 2 | 330 | 137 | |
GenBank mRNA | AY150309.1 | - | 1 - 2 | 158 | 137 |
BC025765.1 | - | 1 - 2 | 158 | 137 | |
BC141821.1 | - | 1 - 2 | 314 | 137 | |
BC142725.1 | - | 1 - 2 | 314 | 137 | |
BX113509.1 | - | 1 - 2 | 266 | 137 |