The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007167.3 | NP_009098.3 | 2 - 3 | 319 | 121 |
GenBank mRNA | AK299293.1 | - | 2 - 3 | 278 | 121 |
AK310703.1 | - | 2 - 3 | 260 | 121 | |
AL389985.1 | - | 2 - 3 | 244 | 121 | |
AL705390.1 | - | 2 - 3 | 272 | 121 | |
AL832911.1 | - | 2 - 3 | 265 | 121 | |
AL833263.1 | - | 2 - 3 | 292 | 121 | |
BC033903.1 | - | 2 - 3 | 301 | 121 | |
BC068534.1 | - | 2 - 3 | 319 | 121 | |
BC107880.1 | - | 2 - 3 | 300 | 121 | |
BC117463.1 | - | 2 - 3 | 187 | 121 | |
BT007117.1 | - | 1 - 2 | 91 | 121 |