Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LACE1
Entrez Gene ID
246269
Gene Name
lactation elevated 1
Gene Aliases
AFG1, c222389
Chromosome Location
Chr.6: 108294874 - 108525784 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001323005.1 NP_001309934.1 4 - 5 722 143
NM_145315.4 NP_660358.2 4 - 5 722 143
XM_005266885.3 XP_005266942.1 5 - 6 2080 143
XM_011535659.2 XP_011533961.1 4 - 5 605 143
XM_011535660.2 XP_011533962.1 4 - 5 605 143
XM_011535661.2 XP_011533963.1 4 - 5 605 143
XM_017010647.1 XP_016866136.1 5 - 6 572 143
XM_017010648.1 XP_016866137.1 3 - 4 174 143
XM_017010649.1 XP_016866138.1 4 - 5 838 143
GenBank mRNA AF520418.1 - 4 - 5 702 143
BC018445.1 - 4 - 5 702 143

View Full Product Details