Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LACE1
Entrez Gene ID
246269
Gene Name
lactation elevated 1
Gene Aliases
AFG1, c222389
Chromosome Location
Chr.6: 108294874 - 108525784 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001323005.1 NP_001309934.1 8 - 9 1095 89
NM_145315.4 NP_660358.2 8 - 9 1095 89
NR_136553.1 4 - 5 658 89
XM_005266885.3 XP_005266942.1 9 - 10 2453 89
XM_011535657.2 XP_011533959.1 7 - 8 876 89
XM_017010647.1 XP_016866136.1 9 - 10 945 89
XM_017010648.1 XP_016866137.1 7 - 8 547 89
XM_017010649.1 XP_016866138.1 8 - 9 1211 89
GenBank mRNA AF520418.1 - 8 - 9 1075 89
BC018445.1 - 8 - 9 1075 89
BI754108.1 - 4 - 5 353 89

View Full Product Details