Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDCD4
Entrez Gene ID
27250
Gene Name
programmed cell death 4 (neoplastic transformation inhibitor)
Gene Aliases
H731
Chromosome Location
Chr.10: 110871795 - 110900006 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199492.1 NP_001186421.1 10 - 11 1456 119
NM_014456.4 NP_055271.2 10 - 11 1498 119
NM_145341.3 NP_663314.1 11 - 12 1582 119
GenBank mRNA AK223623.1 - 10 - 11 1372 119
AK296753.1 - 10 - 11 1323 119
AK312540.1 - 10 - 11 1394 119
AK315295.1 - 11 - 12 1460 119
BC026104.2 - 10 - 11 1365 119
BC031049.1 - 10 - 11 1365 119
BX537500.1 - 8 - 9 5371 119
BX647834.1 - 10 - 11 1340 119
U96628.1 - 10 - 11 1296 119

View Full Product Details