Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCGB1C1
Entrez Gene ID
147199
Gene Name
secretoglobin family 1C member 1
Gene Aliases
RYD5
Chromosome Location
Chr.11: 187881 - 194575 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145651.2 NP_663626.2 1 - 2 74 81
XM_005252804.3 XP_005252861.2 2 - 3 354 81
GenBank mRNA AY026938.1 - 1 - 2 74 81
BC069287.1 - 1 - 2 74 81
BC111427.1 - 1 - 2 73 81
BC111433.1 - 1 - 2 73 81

Gene Symbol
SCGB1C2
Entrez Gene ID
653486
Gene Name
secretoglobin family 1C member 2
Gene Aliases
-
Chromosome Location
Chr.17: 132346 - 139067 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097610.2 NP_001091079.1 1 - 2 81
XM_011535362.2 XP_011533664.1 2 - 3 81

View Full Product Details