Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP4K4
Entrez Gene ID
9448
Gene Name
mitogen-activated protein kinase kinase kinase kinase 4
Gene Aliases
FLH21957, HEL-S-31, HGK, MEKKK4, NIK
Chromosome Location
Chr.2: 101697703 - 101894690 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242560.1 NP_001229489.1 17 - 18 2327 66
NM_145686.3 NP_663719.2 17 - 18 2258 66
NM_145687.3 NP_663720.1 17 - 18 1885 66
XM_005264048.2 XP_005264105.1 18 - 19 2278 66
XM_005264050.3 XP_005264107.1 17 - 18 2116 66
XM_005264057.3 XP_005264114.1 17 - 18 2047 66
XM_005264058.3 XP_005264115.1 17 - 18 2047 66
XM_005264059.3 XP_005264116.1 17 - 18 1954 66
XM_005264062.3 XP_005264119.1 16 - 17 1885 66
XM_005264064.3 XP_005264121.1 16 - 17 1885 66
XM_005264065.3 XP_005264122.1 16 - 17 1792 66
XM_005264068.3 XP_005264125.1 16 - 17 1792 66
XM_005264069.2 XP_005264126.1 17 - 18 1954 66
XM_006712865.2 XP_006712928.1 17 - 18 2023 66
XM_006712868.2 XP_006712931.1 16 - 17 1792 66
XM_006712869.2 XP_006712932.1 16 - 17 1885 66
XM_017005350.1 XP_016860839.1 17 - 18 2116 66
XM_017005352.1 XP_016860841.1 17 - 18 2047 66
XM_017005354.1 XP_016860843.1 17 - 18 2023 66
XM_017005355.1 XP_016860844.1 17 - 18 2023 66
XM_017005356.1 XP_016860845.1 17 - 18 1954 66
XM_017005357.1 XP_016860846.1 17 - 18 1954 66
XM_017005359.1 XP_016860848.1 17 - 18 2116 66
XM_017005361.1 XP_016860850.1 17 - 18 2116 66
XM_017005363.1 XP_016860852.1 16 - 17 1885 66
XM_017005365.1 XP_016860854.1 17 - 18 2047 66
XM_017005366.1 XP_016860855.1 17 - 18 2023 66
XM_017005371.1 XP_016860860.1 16 - 17 1885 66
XM_017005372.1 XP_016860861.1 16 - 17 1792 66
XM_017005373.1 XP_016860862.1 16 - 17 1792 66
XM_017005374.1 XP_016860863.1 16 - 17 1792 66
XM_017005375.1 XP_016860864.1 16 - 17 1792 66
GenBank mRNA AB014587.1 - 15 - 16 1769 66
AK001272.1 - 1 - 2 37 66
AK295610.1 - 15 - 16 1919 66
AK296396.1 - 17 - 18 2023 66
AL137755.1 - 9 - 10 1422 66
AY834276.1 - 12 - 13 1370 66
EU794634.1 - 17 - 18 1935 66

View Full Product Details