Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP4K4
Entrez Gene ID
9448
Gene Name
mitogen-activated protein kinase kinase kinase kinase 4
Gene Aliases
FLH21957, HEL-S-31, HGK, MEKKK4, NIK
Chromosome Location
Chr.2: 101697703 - 101894690 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242559.1 NP_001229488.1 25 - 26 3080 111
NM_001242560.1 NP_001229489.1 26 - 27 3441 111
NM_004834.4 NP_004825.3 25 - 26 3207 111
NM_145686.3 NP_663719.2 26 - 27 3555 111
NM_145687.3 NP_663720.1 26 - 27 2999 111
XM_005264048.2 XP_005264105.1 28 - 29 3584 111
XM_005264050.3 XP_005264107.1 27 - 28 3422 111
XM_005264052.3 XP_005264109.1 27 - 28 3419 111
XM_005264057.3 XP_005264114.1 27 - 28 3353 111
XM_005264058.3 XP_005264115.1 26 - 27 3344 111
XM_005264059.3 XP_005264116.1 27 - 28 3260 111
XM_005264060.3 XP_005264117.1 26 - 27 3248 111
XM_005264062.3 XP_005264119.1 26 - 27 3191 111
XM_005264063.3 XP_005264120.1 26 - 27 3188 111
XM_005264064.3 XP_005264121.1 25 - 26 3182 111
XM_005264065.3 XP_005264122.1 26 - 27 3098 111
XM_005264066.3 XP_005264123.1 26 - 27 3095 111
XM_005264068.3 XP_005264125.1 25 - 26 3089 111
XM_005264069.2 XP_005264126.1 25 - 26 3059 111
XM_005264071.2 XP_005264128.1 24 - 25 2987 111
XM_006712865.2 XP_006712928.1 27 - 28 3329 111
XM_006712868.2 XP_006712931.1 26 - 27 3098 111
XM_006712869.2 XP_006712932.1 25 - 26 2999 111
XM_017005349.1 XP_016860838.1 28 - 29 3488 111
XM_017005350.1 XP_016860839.1 27 - 28 3422 111
XM_017005351.1 XP_016860840.1 26 - 27 3341 111
XM_017005352.1 XP_016860841.1 27 - 28 3353 111
XM_017005353.1 XP_016860842.1 27 - 28 3350 111
XM_017005354.1 XP_016860843.1 27 - 28 3329 111
XM_017005355.1 XP_016860844.1 26 - 27 3320 111
XM_017005356.1 XP_016860845.1 26 - 27 3251 111
XM_017005357.1 XP_016860846.1 27 - 28 3260 111
XM_017005358.1 XP_016860847.1 27 - 28 3257 111
XM_017005359.1 XP_016860848.1 26 - 27 3230 111
XM_017005360.1 XP_016860849.1 26 - 27 3227 111
XM_017005361.1 XP_016860850.1 26 - 27 3230 111
XM_017005362.1 XP_016860851.1 26 - 27 3227 111
XM_017005363.1 XP_016860852.1 26 - 27 3191 111
XM_017005364.1 XP_016860853.1 25 - 26 3179 111
XM_017005365.1 XP_016860854.1 26 - 27 3161 111
XM_017005366.1 XP_016860855.1 26 - 27 3137 111
XM_017005367.1 XP_016860856.1 26 - 27 3134 111
XM_017005368.1 XP_016860857.1 26 - 27 3065 111
XM_017005369.1 XP_016860858.1 25 - 26 2996 111
XM_017005370.1 XP_016860859.1 25 - 26 2996 111
XM_017005371.1 XP_016860860.1 24 - 25 2990 111
XM_017005372.1 XP_016860861.1 25 - 26 2906 111
XM_017005373.1 XP_016860862.1 24 - 25 2897 111
XM_017005374.1 XP_016860863.1 25 - 26 2906 111
XM_017005375.1 XP_016860864.1 24 - 25 2897 111
GenBank mRNA AB013385.1 - 9 - 10 987 111
AB014587.1 - 24 - 25 2883 111
AF096300.1 - 25 - 26 2908 111
AK001272.1 - 9 - 10 1142 111
AK025610.1 - 13 - 14 1756 111
AK074592.1 - 5 - 6 748 111
AK295610.1 - 25 - 26 3225 111
AK296396.1 - 26 - 27 3137 111
AL137755.1 - 18 - 19 2536 111
AY212247.1 - 25 - 26 3075 111
EU794634.1 - 26 - 27 3049 111

View Full Product Details