The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105513.2 | NP_001098983.2 | 12 - 13 | 1730 | 70 |
NM_145755.2 | NP_665698.2 | 13 - 14 | 1874 | 70 | |
XM_005264921.4 | XP_005264978.1 | 13 - 14 | 1803 | 70 | |
XM_005264922.4 | XP_005264979.1 | 13 - 14 | 1803 | 70 | |
XM_005264923.3 | XP_005264980.1 | 13 - 14 | 1803 | 70 | |
XM_005264924.4 | XP_005264981.1 | 13 - 14 | 1803 | 70 | |
XM_005264925.4 | XP_005264982.1 | 12 - 13 | 1680 | 70 | |
XM_005264926.4 | XP_005264983.1 | 13 - 14 | 1803 | 70 | |
XM_005264927.4 | XP_005264984.1 | 9 - 10 | 1257 | 70 | |
XM_006713011.3 | XP_006713074.1 | 13 - 14 | 1803 | 70 | |
XM_011533447.2 | XP_011531749.1 | 12 - 13 | 1704 | 70 | |
XM_011533449.1 | XP_011531751.1 | 13 - 14 | 1803 | 70 | |
XM_017005843.1 | XP_016861332.1 | 13 - 14 | 1803 | 70 | |
GenBank mRNA | AJ487015.1 | - | 13 - 14 | 1836 | 70 |
AK093313.1 | - | 3 - 4 | 414 | 70 | |
AK302408.1 | - | 12 - 13 | 1713 | 70 | |
AK302465.1 | - | 12 - 13 | 1584 | 70 | |
BC129948.1 | - | 13 - 14 | 1785 | 70 |