The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145913.3 | NP_666018.3 | 8 - 9 | 1441 | 105 |
XM_017018910.1 | XP_016874399.1 | 8 - 9 | 1477 | 105 | |
GenBank mRNA | AF536216.1 | - | 8 - 9 | 1414 | 105 |
AK075263.1 | - | 8 - 9 | 1099 | 105 | |
AK313788.1 | - | 8 - 9 | 1410 | 105 | |
AY081220.1 | - | 8 - 9 | 1415 | 105 | |
BC110492.2 | - | 8 - 9 | 1076 | 105 |