Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WHRN
Entrez Gene ID
25861
Gene Name
whirlin
Gene Aliases
CIP98, DFNB31, PDZD7B, USH2D, WI
Chromosome Location
Chr.9: 114402078 - 114505509 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083885.2 NP_001077354.2 7 - 8 1118 95
NM_001173425.1 NP_001166896.1 7 - 8 2282 95
NM_015404.3 NP_056219.3 7 - 8 2282 95
GenBank mRNA AB040959.1 - 7 - 8 1796 95
AK022854.1 - 3 - 4 996 95
AL110228.1 - 7 - 8 1076 95
BC014524.1 - - 416 95
BC136416.1 - 7 - 8 2031 95
BC142614.1 - 7 - 8 2282 95
BC142684.1 - 7 - 8 2257 95

View Full Product Details