Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 24476291-24476823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM7
Entrez Gene ID
8756
Gene Name
ADAM metallopeptidase domain 7
Gene Aliases
ADAM 7, ADAM-7, EAPI, GP-83, GP83
UniGene
Hs.116147
Gene Chromosome Location
Chr.8: 24440996-24509565 on Build GRCh38
Transcripts
NM_003817.3 XM_017013943.1 NR_125808.1

Gene Symbol
LOC101929294
Entrez Gene ID
101929294
Gene Name
uncharacterized LOC101929294
Gene Aliases
-
UniGene
Hs.637673
Gene Chromosome Location
Chr.8: 24295814-24548618 on Build GRCh38
Transcripts
NM_003817.3 XM_017013943.1 NR_125808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5997910 c.651T>C p.H217H Chr.8: 24476450-24476450 on Build GRCh38
1684881 c.673C>T p.R225* Chr.8: 24476472-24476472 on Build GRCh38
4977120 c.638G>T p.R213L Chr.8: 24476437-24476437 on Build GRCh38
5907467 c.634-4C>T p.? Chr.8: 24476429-24476429 on Build GRCh38
1456204 c.655C>T p.H219Y Chr.8: 24476454-24476454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150617851 Chr.8: 24476486-24476486 on Build GRCh38
rs200192082 Chr.8: 24476544-24476544 on Build GRCh38
rs895181357 Chr.8: 24476367-24476367 on Build GRCh38
rs150996912 Chr.8: 24476456-24476456 on Build GRCh38
rs372347679 Chr.8: 24476413-24476413 on Build GRCh38
rs996737965 Chr.8: 24476674-24476674 on Build GRCh38
rs752292825 Chr.8: 24476407-24476407 on Build GRCh38
rs953768139 Chr.8: 24476481-24476481 on Build GRCh38
rs945039579 Chr.8: 24476409-24476409 on Build GRCh38
rs371616825 Chr.8: 24476518-24476518 on Build GRCh38
rs566385438 Chr.8: 24476372-24476372 on Build GRCh38
rs781280181 Chr.8: 24476537-24476537 on Build GRCh38
rs559549236 Chr.8: 24476601-24476602 on Build GRCh38
rs902043940 Chr.8: 24476654-24476654 on Build GRCh38
rs895036561 Chr.8: 24476570-24476570 on Build GRCh38
rs1055446736 Chr.8: 24476334-24476334 on Build GRCh38
rs754624973 Chr.8: 24476403-24476403 on Build GRCh38
rs565665566 Chr.8: 24476364-24476364 on Build GRCh38
rs746112199 Chr.8: 24476437-24476437 on Build GRCh38
rs930678463 Chr.8: 24476575-24476575 on Build GRCh38
rs374050534 Chr.8: 24476560-24476560 on Build GRCh38
rs555462518 Chr.8: 24476565-24476565 on Build GRCh38
rs574902650 Chr.8: 24476587-24476587 on Build GRCh38
rs1003420316 Chr.8: 24476332-24476332 on Build GRCh38
rs140840518 Chr.8: 24476642-24476642 on Build GRCh38
rs1055079581 Chr.8: 24476569-24476569 on Build GRCh38
rs372611510 Chr.8: 24476548-24476548 on Build GRCh38
rs539044011 Chr.8: 24476381-24476381 on Build GRCh38
rs763376612 Chr.8: 24476489-24476489 on Build GRCh38
rs977843285 Chr.8: 24476778-24476778 on Build GRCh38
rs780315494 Chr.8: 24476529-24476529 on Build GRCh38
rs771416007 Chr.8: 24476555-24476555 on Build GRCh38
rs1029653786 Chr.8: 24476480-24476480 on Build GRCh38
rs759469139 Chr.8: 24476582-24476582 on Build GRCh38
rs184696574 Chr.8: 24476615-24476615 on Build GRCh38
rs773407673 Chr.8: 24476406-24476406 on Build GRCh38
rs989112845 Chr.8: 24476711-24476711 on Build GRCh38
rs773667941 Chr.8: 24476479-24476479 on Build GRCh38
rs777076478 Chr.8: 24476472-24476472 on Build GRCh38
rs141868243 Chr.8: 24476478-24476478 on Build GRCh38
rs763743819 Chr.8: 24476517-24476518 on Build GRCh38
rs914968418 Chr.8: 24476650-24476650 on Build GRCh38
rs12549198 Chr.8: 24476756-24476756 on Build GRCh38
rs770065361 Chr.8: 24476373-24476373 on Build GRCh38
rs777628131 Chr.8: 24476416-24476416 on Build GRCh38
rs140756552 Chr.8: 24476500-24476500 on Build GRCh38
rs766093123 Chr.8: 24476498-24476498 on Build GRCh38
rs1019038509 Chr.8: 24476643-24476643 on Build GRCh38
rs772244085 Chr.8: 24476444-24476444 on Build GRCh38
rs536701350 Chr.8: 24476530-24476530 on Build GRCh38
rs753731929 Chr.8: 24476515-24476515 on Build GRCh38
rs1049061249 Chr.8: 24476576-24476576 on Build GRCh38
rs775659869 Chr.8: 24476451-24476451 on Build GRCh38
rs397952282 Chr.8: 24476342-24476343 on Build GRCh38
rs767163897 Chr.8: 24476404-24476404 on Build GRCh38
rs60628806 Chr.8: 24476335-24476336 on Build GRCh38
rs6983897 Chr.8: 24476387-24476387 on Build GRCh38
rs369319361 Chr.8: 24476463-24476463 on Build GRCh38
rs762516782 Chr.8: 24476414-24476416 on Build GRCh38
rs768771407 Chr.8: 24476457-24476457 on Build GRCh38
rs763467237 Chr.8: 24476551-24476551 on Build GRCh38
rs1029272750 Chr.8: 24476689-24476689 on Build GRCh38
rs148539948 Chr.8: 24476436-24476436 on Build GRCh38
rs745704021 Chr.8: 24476520-24476520 on Build GRCh38
rs977895765 Chr.8: 24476534-24476534 on Build GRCh38
rs17052036 Chr.8: 24476758-24476758 on Build GRCh38
rs375036002 Chr.8: 24476526-24476526 on Build GRCh38
rs906466257 Chr.8: 24476320-24476320 on Build GRCh38
rs547272948 Chr.8: 24476363-24476363 on Build GRCh38
rs957834565 Chr.8: 24476697-24476697 on Build GRCh38
rs749243622 Chr.8: 24476417-24476417 on Build GRCh38
rs757007216 Chr.8: 24476427-24476427 on Build GRCh38
rs62500004 Chr.8: 24476687-24476687 on Build GRCh38
rs1016995032 Chr.8: 24476512-24476512 on Build GRCh38
rs768626890 Chr.8: 24476535-24476535 on Build GRCh38
rs767332661 Chr.8: 24476628-24476628 on Build GRCh38
rs774185650 Chr.8: 24476550-24476550 on Build GRCh38
rs761875325 Chr.8: 24476448-24476448 on Build GRCh38
rs767325259 Chr.8: 24476494-24476494 on Build GRCh38
rs1005068187 Chr.8: 24476433-24476433 on Build GRCh38

View Full Product Details