Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 26408038-26408533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNIP3L
Entrez Gene ID
665
Gene Name
BCL2 interacting protein 3 like
Gene Aliases
BNIP3a, NIX
UniGene
Hs.131226
Gene Chromosome Location
Chr.8: 26383007-26413128 on Build GRCh38
Transcripts
NM_004331.2 XM_005273617.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2787451 c.611+10G>A p.? Chr.8: 26408386-26408386 on Build GRCh38
6239056 c.471C>T p.H157H Chr.8: 26408236-26408236 on Build GRCh38
5998001 c.436A>G p.S146G Chr.8: 26408078-26408078 on Build GRCh38
2787447 c.453T>C p.I151I Chr.8: 26408095-26408095 on Build GRCh38
6206961 c.407A>G p.K136R Chr.8: 26408049-26408049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918195494 Chr.8: 26408270-26408270 on Build GRCh38
rs901804164 Chr.8: 26408458-26408458 on Build GRCh38
rs866392776 Chr.8: 26408063-26408063 on Build GRCh38
rs202020745 Chr.8: 26408392-26408392 on Build GRCh38
rs200534833 Chr.8: 26408086-26408086 on Build GRCh38
rs771290000 Chr.8: 26408298-26408298 on Build GRCh38
rs150321619 Chr.8: 26408116-26408116 on Build GRCh38
rs14994 Chr.8: 26408311-26408311 on Build GRCh38
rs758200966 Chr.8: 26408117-26408117 on Build GRCh38
rs369015422 Chr.8: 26408358-26408358 on Build GRCh38
rs777644458 Chr.8: 26408293-26408293 on Build GRCh38
rs556579869 Chr.8: 26408491-26408491 on Build GRCh38
rs760342858 Chr.8: 26408178-26408178 on Build GRCh38
rs774517642 Chr.8: 26408153-26408153 on Build GRCh38
rs570755475 Chr.8: 26408442-26408442 on Build GRCh38
rs770467255 Chr.8: 26408148-26408148 on Build GRCh38
rs774809502 Chr.8: 26408300-26408300 on Build GRCh38
rs369179592 Chr.8: 26408116-26408117 on Build GRCh38
rs369241811 Chr.8: 26408395-26408395 on Build GRCh38
rs958900366 Chr.8: 26408480-26408480 on Build GRCh38
rs751267986 Chr.8: 26408244-26408244 on Build GRCh38
rs564581632 Chr.8: 26408172-26408172 on Build GRCh38
rs113593460 Chr.8: 26408151-26408151 on Build GRCh38
rs373938692 Chr.8: 26408312-26408312 on Build GRCh38
rs575926839 Chr.8: 26408220-26408220 on Build GRCh38
rs777680555 Chr.8: 26408201-26408202 on Build GRCh38
rs753386018 Chr.8: 26408202-26408202 on Build GRCh38
rs570031960 Chr.8: 26408501-26408501 on Build GRCh38
rs747260723 Chr.8: 26408139-26408139 on Build GRCh38
rs757216151 Chr.8: 26408093-26408093 on Build GRCh38
rs746836011 Chr.8: 26408252-26408253 on Build GRCh38
rs370470878 Chr.8: 26408385-26408385 on Build GRCh38
rs751442664 Chr.8: 26408386-26408386 on Build GRCh38
rs368261648 Chr.8: 26408147-26408147 on Build GRCh38
rs867912249 Chr.8: 26408354-26408354 on Build GRCh38
rs138001749 Chr.8: 26408152-26408152 on Build GRCh38
rs532017041 Chr.8: 26408222-26408222 on Build GRCh38
rs761289218 Chr.8: 26408208-26408208 on Build GRCh38
rs140691550 Chr.8: 26408288-26408288 on Build GRCh38
rs756229023 Chr.8: 26408407-26408407 on Build GRCh38
rs776876975 Chr.8: 26408144-26408144 on Build GRCh38
rs764809129 Chr.8: 26408328-26408328 on Build GRCh38
rs932465094 Chr.8: 26408287-26408287 on Build GRCh38
rs778789585 Chr.8: 26408095-26408095 on Build GRCh38
rs763603270 Chr.8: 26408067-26408067 on Build GRCh38
rs749811787 Chr.8: 26408297-26408297 on Build GRCh38
rs1036247570 Chr.8: 26408100-26408100 on Build GRCh38
rs139026759 Chr.8: 26408236-26408236 on Build GRCh38
rs150078014 Chr.8: 26408065-26408065 on Build GRCh38
rs763695305 Chr.8: 26408189-26408189 on Build GRCh38
rs771507540 Chr.8: 26408423-26408423 on Build GRCh38
rs539869098 Chr.8: 26408469-26408469 on Build GRCh38
rs759141869 Chr.8: 26408303-26408303 on Build GRCh38
rs749845213 Chr.8: 26408347-26408347 on Build GRCh38
rs766190462 Chr.8: 26408377-26408377 on Build GRCh38
rs572517609 Chr.8: 26408135-26408135 on Build GRCh38
rs757084399 Chr.8: 26408386-26408386 on Build GRCh38
rs748289954 Chr.8: 26408289-26408289 on Build GRCh38
rs749287064 Chr.8: 26408421-26408421 on Build GRCh38
rs901751908 Chr.8: 26408113-26408114 on Build GRCh38
rs755286076 Chr.8: 26408253-26408253 on Build GRCh38
rs991166974 Chr.8: 26408133-26408133 on Build GRCh38
rs943231585 Chr.8: 26408400-26408400 on Build GRCh38
rs376715919 Chr.8: 26408241-26408241 on Build GRCh38
rs200081637 Chr.8: 26408314-26408314 on Build GRCh38
rs908467401 Chr.8: 26408166-26408166 on Build GRCh38
rs149504745 Chr.8: 26408409-26408409 on Build GRCh38
rs943926993 Chr.8: 26408182-26408182 on Build GRCh38
rs9644114 Chr.8: 26408391-26408391 on Build GRCh38
rs1021374721 Chr.8: 26408503-26408503 on Build GRCh38

View Full Product Details