Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 26412512-26413013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNIP3L
Entrez Gene ID
665
Gene Name
BCL2 interacting protein 3 like
Gene Aliases
BNIP3a, NIX
UniGene
Hs.131226
Gene Chromosome Location
Chr.8: 26383007-26413128 on Build GRCh38
Transcripts
NM_004331.2 XM_005273617.3

SNP Detail

SNP ID Chromosome Location
rs1014150480 Chr.8: 26412977-26412977 on Build GRCh38
rs76206561 Chr.8: 26412683-26412683 on Build GRCh38
rs536086157 Chr.8: 26412895-26412895 on Build GRCh38
rs547148115 Chr.8: 26412590-26412590 on Build GRCh38
rs968567976 Chr.8: 26412579-26412579 on Build GRCh38
rs767320386 Chr.8: 26412893-26412893 on Build GRCh38
rs746760106 Chr.8: 26412650-26412650 on Build GRCh38
rs747657735 Chr.8: 26412742-26412742 on Build GRCh38
rs761747030 Chr.8: 26412824-26412824 on Build GRCh38
rs567352386 Chr.8: 26412894-26412894 on Build GRCh38
rs552877172 Chr.8: 26412681-26412681 on Build GRCh38
rs779840733 Chr.8: 26412615-26412615 on Build GRCh38
rs993243806 Chr.8: 26412626-26412626 on Build GRCh38
rs768333001 Chr.8: 26412657-26412657 on Build GRCh38
rs537012827 Chr.8: 26412790-26412790 on Build GRCh38
rs532874670 Chr.8: 26412630-26412630 on Build GRCh38
rs930837833 Chr.8: 26412740-26412740 on Build GRCh38
rs758204368 Chr.8: 26412594-26412594 on Build GRCh38
rs186863667 Chr.8: 26412603-26412603 on Build GRCh38
rs571270115 Chr.8: 26412778-26412778 on Build GRCh38
rs530274894 Chr.8: 26412589-26412589 on Build GRCh38
rs972095360 Chr.8: 26412661-26412661 on Build GRCh38
rs920668801 Chr.8: 26412666-26412666 on Build GRCh38
rs1020141922 Chr.8: 26412573-26412573 on Build GRCh38
rs1000469716 Chr.8: 26412941-26412941 on Build GRCh38
rs1041126399 Chr.8: 26412821-26412821 on Build GRCh38
rs894383393 Chr.8: 26412970-26412970 on Build GRCh38
rs904268825 Chr.8: 26412913-26412913 on Build GRCh38
rs1034149788 Chr.8: 26412950-26412950 on Build GRCh38
rs1050615331 Chr.8: 26412746-26412747 on Build GRCh38
rs1000014681 Chr.8: 26412582-26412582 on Build GRCh38
rs553118428 Chr.8: 26412903-26412903 on Build GRCh38
rs900810197 Chr.8: 26412845-26412845 on Build GRCh38
rs550444962 Chr.8: 26412851-26412851 on Build GRCh38
rs561422332 Chr.8: 26412562-26412562 on Build GRCh38
rs944943172 Chr.8: 26412804-26412804 on Build GRCh38
rs917210953 Chr.8: 26412635-26412635 on Build GRCh38
rs369042805 Chr.8: 26412915-26412915 on Build GRCh38

View Full Product Details