Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 26412854-26413350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNIP3L
Entrez Gene ID
665
Gene Name
BCL2 interacting protein 3 like
Gene Aliases
BNIP3a, NIX
UniGene
Hs.131226
Gene Chromosome Location
Chr.8: 26383007-26413128 on Build GRCh38
Transcripts
NM_004331.2 XM_005273617.3

SNP Detail

SNP ID Chromosome Location
rs567352386 Chr.8: 26412894-26412894 on Build GRCh38
rs1014493265 Chr.8: 26413214-26413214 on Build GRCh38
rs962049150 Chr.8: 26413027-26413027 on Build GRCh38
rs897745512 Chr.8: 26413240-26413240 on Build GRCh38
rs958126673 Chr.8: 26413265-26413265 on Build GRCh38
rs572672935 Chr.8: 26413052-26413052 on Build GRCh38
rs1027577914 Chr.8: 26413267-26413267 on Build GRCh38
rs1000469716 Chr.8: 26412941-26412941 on Build GRCh38
rs142844378 Chr.8: 26413205-26413205 on Build GRCh38
rs1056054478 Chr.8: 26413208-26413208 on Build GRCh38
rs148509730 Chr.8: 26413126-26413126 on Build GRCh38
rs894383393 Chr.8: 26412970-26412970 on Build GRCh38
rs538547632 Chr.8: 26413053-26413053 on Build GRCh38
rs904268825 Chr.8: 26412913-26412913 on Build GRCh38
rs558021369 Chr.8: 26413213-26413213 on Build GRCh38
rs1034149788 Chr.8: 26412950-26412950 on Build GRCh38
rs1024202170 Chr.8: 26413239-26413239 on Build GRCh38
rs910682290 Chr.8: 26413171-26413176 on Build GRCh38
rs750258874 Chr.8: 26413038-26413038 on Build GRCh38
rs553118428 Chr.8: 26412903-26412903 on Build GRCh38
rs986203049 Chr.8: 26413287-26413287 on Build GRCh38
rs966258998 Chr.8: 26413289-26413289 on Build GRCh38
rs147706417 Chr.8: 26413303-26413303 on Build GRCh38
rs767755497 Chr.8: 26413235-26413236 on Build GRCh38
rs954618719 Chr.8: 26413191-26413191 on Build GRCh38
rs369042805 Chr.8: 26412915-26412915 on Build GRCh38
rs1002355594 Chr.8: 26413162-26413162 on Build GRCh38
rs1014150480 Chr.8: 26412977-26412977 on Build GRCh38
rs769329556 Chr.8: 26413004-26413004 on Build GRCh38
rs536086157 Chr.8: 26412895-26412895 on Build GRCh38
rs986712131 Chr.8: 26413154-26413154 on Build GRCh38
rs1046558922 Chr.8: 26413128-26413128 on Build GRCh38
rs767320386 Chr.8: 26412893-26412893 on Build GRCh38
rs988551426 Chr.8: 26413269-26413269 on Build GRCh38
rs945733566 Chr.8: 26413194-26413194 on Build GRCh38
rs187087918 Chr.8: 26413304-26413304 on Build GRCh38
rs183413558 Chr.8: 26413195-26413195 on Build GRCh38
rs1017530831 Chr.8: 26413204-26413204 on Build GRCh38
rs1024275844 Chr.8: 26413016-26413016 on Build GRCh38
rs913873599 Chr.8: 26413288-26413288 on Build GRCh38
rs993587218 Chr.8: 26413180-26413180 on Build GRCh38
rs755844659 Chr.8: 26413188-26413188 on Build GRCh38
rs1027735885 Chr.8: 26413102-26413102 on Build GRCh38
rs952231340 Chr.8: 26413274-26413274 on Build GRCh38
rs976244683 Chr.8: 26413292-26413292 on Build GRCh38
rs191255981 Chr.8: 26413176-26413176 on Build GRCh38
rs905405951 Chr.8: 26413150-26413150 on Build GRCh38
rs575305275 Chr.8: 26413065-26413065 on Build GRCh38
rs774785224 Chr.8: 26413170-26413170 on Build GRCh38
rs776017072 Chr.8: 26413229-26413229 on Build GRCh38

View Full Product Details