The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175732.2 | NP_783859.1 | 2 - 3 | 444 | 68 |
GenBank mRNA | BC014048.1 | - | 1 - 2 | 90 | 68 |
BC020242.1 | - | 2 - 3 | 273 | 68 | |
DA476984.1 | - | 2 - 3 | 300 | 68 | |
DR004030.1 | - | 2 - 3 | 302 | 68 |