The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152501.4 | NP_689714.2 | 3 - 4 | 656 | 81 |
NM_198928.4 | NP_945146.1 | 3 - 4 | 629 | 81 | |
NM_198929.4 | NP_945147.1 | 3 - 4 | 656 | 81 | |
NM_198930.3 | NP_945148.1 | 3 - 4 | 629 | 81 | |
XM_005244930.1 | XP_005244987.1 | 3 - 4 | 656 | 81 | |
XM_011509242.2 | XP_011507544.1 | 3 - 4 | 656 | 81 | |
XM_011509243.2 | XP_011507545.1 | 3 - 4 | 656 | 81 | |
XM_017000463.1 | XP_016855952.1 | 3 - 4 | 656 | 81 | |
GenBank mRNA | AK308350.1 | - | 3 - 4 | 635 | 82 |
AY185344.1 | - | 3 - 4 | 643 | 81 | |
AY185345.1 | - | 3 - 4 | 616 | 81 | |
AY185346.1 | - | 3 - 4 | 643 | 81 | |
AY185347.1 | - | 3 - 4 | 616 | 81 | |
BC020822.1 | - | 3 - 4 | 635 | 81 | |
BC090944.1 | - | 3 - 4 | 638 | 81 | |
BC139741.1 | - | 3 - 4 | 583 | 81 |