The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009936.2 | NP_001009936.1 | 1 - 2 | 235 | 78 |
NM_001286843.1 | NP_001273772.1 | 1 - 2 | 235 | 78 | |
NM_015651.2 | NP_056466.1 | 1 - 2 | 235 | 78 | |
GenBank mRNA | AK122744.1 | - | 1 - 2 | 95 | 78 |
AK303433.1 | - | 1 - 2 | 125 | 78 | |
BC022374.1 | - | 1 - 2 | 95 | 78 | |
BC108663.1 | - | 1 - 2 | 202 | 78 | |
BM784783.1 | - | 1 - 2 | 235 | 78 | |
BM784790.1 | - | 1 - 2 | 235 | 78 |