Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCDN
Entrez Gene ID
23154
Gene Name
neurochondrin
Gene Aliases
-
Chromosome Location
Chr.1: 35557792 - 35566779 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014839.1 NP_001014839.1 4 - 5 1270 91
NM_001014841.1 NP_001014841.1 3 - 4 1202 91
NM_014284.2 NP_055099.1 3 - 4 1542 91
GenBank mRNA AB011179.1 - 3 - 4 1150 91
AB018739.2 - 4 - 5 1254 91
AB018740.2 - 3 - 4 1186 91
BC024592.1 - 3 - 4 1516 91

View Full Product Details