Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOP9
Entrez Gene ID
161424
Gene Name
NOP9 nucleolar protein
Gene Aliases
C14orf21
Chromosome Location
Chr.14: 24299854 - 24309126 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286367.1 NP_001273296.1 2 - 3 798 73
NM_174913.2 NP_777573.1 2 - 3 798 73
GenBank mRNA AB095941.1 - 2 - 3 790 73
BC025332.1 - 2 - 3 776 73
BX248018.1 - 2 - 3 760 73
BX647311.1 - 2 - 3 765 73

View Full Product Details