The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052924.2 | NP_443156.2 | 5 - 6 | 589 | 86 |
XM_005250772.3 | XP_005250829.1 | 6 - 7 | 775 | 86 | |
XM_005250773.3 | XP_005250830.1 | 6 - 7 | 775 | 86 | |
XM_006716502.3 | XP_006716565.1 | 6 - 7 | 775 | 86 | |
XM_011516817.2 | XP_011515119.1 | 6 - 7 | 775 | 86 | |
XM_011516818.1 | XP_011515120.1 | 6 - 7 | 1432 | 86 | |
XM_011516819.2 | XP_011515121.1 | 5 - 6 | 616 | 86 | |
XM_011516820.2 | XP_011515122.1 | 5 - 6 | 959 | 86 | |
XM_011516821.2 | XP_011515123.1 | 5 - 6 | 963 | 86 | |
XM_011516822.2 | XP_011515124.1 | 5 - 6 | 1108 | 86 | |
XM_011516823.2 | XP_011515125.1 | 6 - 7 | 776 | 86 | |
XM_017013013.1 | XP_016868502.1 | 5 - 6 | 647 | 86 | |
XM_017013014.1 | XP_016868503.1 | 6 - 7 | 776 | 86 | |
XM_017013015.1 | XP_016868504.1 | 5 - 6 | 1107 | 86 | |
XM_017013016.1 | XP_016868505.1 | 6 - 7 | 775 | 86 | |
GenBank mRNA | AB067516.1 | - | 5 - 6 | 447 | 86 |
AY082588.1 | - | 5 - 6 | 456 | 86 | |
BC025767.1 | - | 5 - 6 | 557 | 86 | |
BX341691.2 | - | 5 - 6 | 597 | 86 |