Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WFDC3
Entrez Gene ID
140686
Gene Name
WAP four-disulfide core domain 3
Gene Aliases
WAP14, dJ447F3.3
Chromosome Location
Chr.20: 45708860 - 45791908 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC026014.1 - 1 - 2 72 61
BC102013.1 - 1 - 2 72 61
BX091378.1 - 1 - 2 44 61

View Full Product Details