The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190456.1 | NP_001177385.1 | 5 - 6 | 704 | 80 |
NM_001190457.1 | NP_001177386.1 | 5 - 6 | 983 | 80 | |
NM_001324014.1 | NP_001310943.1 | 6 - 7 | 915 | 80 | |
NM_001324015.1 | NP_001310944.1 | 5 - 6 | 716 | 80 | |
NM_006091.4 | NP_006082.3 | 5 - 6 | 938 | 80 | |
GenBank mRNA | AB010098.1 | - | 5 - 6 | 671 | 80 |
AB023142.1 | - | 4 - 5 | 642 | 80 | |
AK289678.1 | - | 5 - 6 | 983 | 80 | |
BC026335.1 | - | 5 - 6 | 654 | 80 |