The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018420.2 | NP_060890.2 | 3 - 4 | 567 | 116 |
XM_005271004.3 | XP_005271061.1 | 3 - 4 | 528 | 116 | |
GenBank mRNA | AK125072.1 | - | 1 - 2 | 135 | 115 |
AY145501.1 | - | 3 - 4 | 437 | 116 | |
AY358258.1 | - | 3 - 4 | 451 | 116 | |
BC026358.1 | - | 3 - 4 | 566 | 116 | |
KU178698.1 | - | 3 - 4 | 401 | 116 | |
KU178699.1 | - | 3 - 4 | 401 | 116 | |
KU178700.1 | - | 3 - 4 | 401 | 116 | |
KU178701.1 | - | 2 - 3 | 188 | 116 | |
KU178702.1 | - | 3 - 4 | 401 | 116 | |
KU178703.1 | - | 2 - 3 | 188 | 116 |