Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GIMAP7
Entrez Gene ID
168537
Gene Name
GTPase, IMAP family member 7
Gene Aliases
IAN7, hIAN7
Chromosome Location
Chr.7: 150514857 - 150521073 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153236.3 NP_694968.1 1 - 2 93 82
GenBank mRNA BC006010.1 - 1 - 2 81 82
BC027613.1 - 1 - 2 82 82

View Full Product Details