Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDP2
Entrez Gene ID
57546
Gene Name
pyruvate dehyrogenase phosphatase catalytic subunit 2
Gene Aliases
PPM2B, PPM2C2
Chromosome Location
Chr.16: 66880397 - 66891101 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020786.2 NP_065837.1 1 - 2 160 92
GenBank mRNA AB037769.1 - 1 - 2 107 92
AK292539.1 - 1 - 2 160 92
BC028030.1 - 1 - 2 95 92

View Full Product Details