Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIER2
Entrez Gene ID
54531
Gene Name
MIER family member 2
Gene Aliases
KIAA1193, Mi-er2
Chromosome Location
Chr.19: 305573 - 344793 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017550.1 NP_060020.1 5 - 6 502 65
XM_005259582.4 XP_005259639.1 5 - 6 1086 65
XM_006722769.3 XP_006722832.1 5 - 6 1090 65
XM_011528072.2 XP_011526374.1 5 - 6 1088 65
XM_011528073.1 XP_011526375.1 5 - 6 504 65
XM_011528074.2 XP_011526376.1 5 - 6 1092 65
XM_011528075.2 XP_011526377.1 5 - 6 1609 65
XM_011528077.2 XP_011526379.1 6 - 7 3450 65
XM_011528081.2 XP_011526383.1 3 - 4 346 65
XM_011528082.2 XP_011526384.1 3 - 4 277 65
GenBank mRNA AB033019.1 - - 490 65
BC028203.1 - 5 - 6 502 65

View Full Product Details