Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 179099596-179100099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABL2
Entrez Gene ID
27
Gene Name
ABL proto-oncogene 2, non-receptor tyrosine kinase
Gene Aliases
ABLL, ARG
UniGene
Hs.159472
Gene Chromosome Location
Chr.1: 179099327-179229693 on Build GRCh38
Transcripts
NM_001136000.2 NM_001168236.1 NM_001168237.1 NM_001168238.1 NM_001168239.1 NM_005158.4 NM_007314.3

SNP Detail

SNP ID Chromosome Location
rs1038642846 Chr.1: 179099758-179099758 on Build GRCh38
rs529178364 Chr.1: 179099706-179099706 on Build GRCh38
rs1008940220 Chr.1: 179099874-179099874 on Build GRCh38
rs969570337 Chr.1: 179100037-179100037 on Build GRCh38
rs80122319 Chr.1: 179100071-179100071 on Build GRCh38
rs955544707 Chr.1: 179099970-179099970 on Build GRCh38
rs948975812 Chr.1: 179099904-179099904 on Build GRCh38
rs990227505 Chr.1: 179100019-179100019 on Build GRCh38
rs1047731828 Chr.1: 179099855-179099855 on Build GRCh38
rs777983120 Chr.1: 179099632-179099633 on Build GRCh38
rs923619555 Chr.1: 179099682-179099682 on Build GRCh38
rs886449303 Chr.1: 179099862-179099862 on Build GRCh38
rs991822146 Chr.1: 179100006-179100006 on Build GRCh38
rs572950499 Chr.1: 179099885-179099885 on Build GRCh38
rs747512627 Chr.1: 179099636-179099636 on Build GRCh38
rs568742526 Chr.1: 179099677-179099677 on Build GRCh38
rs575517569 Chr.1: 179099973-179099973 on Build GRCh38
rs544207369 Chr.1: 179100012-179100012 on Build GRCh38
rs1022532014 Chr.1: 179099675-179099675 on Build GRCh38
rs760732876 Chr.1: 179099643-179099643 on Build GRCh38
rs759979676 Chr.1: 179099891-179099891 on Build GRCh38
rs990280000 Chr.1: 179099852-179099852 on Build GRCh38
rs960409125 Chr.1: 179099686-179099686 on Build GRCh38
rs554914928 Chr.1: 179099633-179099633 on Build GRCh38
rs537013461 Chr.1: 179099700-179099700 on Build GRCh38
rs955441446 Chr.1: 179099789-179099789 on Build GRCh38
rs545879249 Chr.1: 179099728-179099728 on Build GRCh38
rs9662858 Chr.1: 179099716-179099716 on Build GRCh38
rs534991975 Chr.1: 179099853-179099853 on Build GRCh38
rs960361204 Chr.1: 179099974-179099974 on Build GRCh38
rs916372353 Chr.1: 179099720-179099720 on Build GRCh38
rs919837815 Chr.1: 179099792-179099792 on Build GRCh38
rs924728810 Chr.1: 179099837-179099837 on Build GRCh38
rs977759661 Chr.1: 179099681-179099681 on Build GRCh38
rs565725020 Chr.1: 179099908-179099908 on Build GRCh38
rs753736413 Chr.1: 179099940-179099940 on Build GRCh38
rs1005352782 Chr.1: 179099951-179099951 on Build GRCh38
rs996339781 Chr.1: 179099934-179099934 on Build GRCh38
rs929902310 Chr.1: 179099793-179099793 on Build GRCh38
rs1040473798 Chr.1: 179099896-179099896 on Build GRCh38
rs970272750 Chr.1: 179100045-179100045 on Build GRCh38
rs957458966 Chr.1: 179100009-179100009 on Build GRCh38
rs939025700 Chr.1: 179099935-179099935 on Build GRCh38
rs757089820 Chr.1: 179099968-179099968 on Build GRCh38
rs957510390 Chr.1: 179099838-179099838 on Build GRCh38
rs141905964 Chr.1: 179099962-179099962 on Build GRCh38
rs375812293 Chr.1: 179099991-179099991 on Build GRCh38
rs771449857 Chr.1: 179099672-179099672 on Build GRCh38
rs561793550 Chr.1: 179099967-179099967 on Build GRCh38
rs114406329 Chr.1: 179099727-179099727 on Build GRCh38
rs998830361 Chr.1: 179099990-179099990 on Build GRCh38
rs751177887 Chr.1: 179099914-179099915 on Build GRCh38
rs1023330062 Chr.1: 179100007-179100007 on Build GRCh38
rs542011194 Chr.1: 179099956-179099956 on Build GRCh38
rs779471460 Chr.1: 179099973-179099974 on Build GRCh38

View Full Product Details