Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 179101338-179101887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABL2
Entrez Gene ID
27
Gene Name
ABL proto-oncogene 2, non-receptor tyrosine kinase
Gene Aliases
ABLL, ARG
UniGene
Hs.159472
Gene Chromosome Location
Chr.1: 179099327-179229693 on Build GRCh38
Transcripts
NM_001136000.2 NM_001168236.1 NM_001168237.1 NM_001168238.1 NM_001168239.1 NM_005158.4 NM_007314.3

SNP Detail

SNP ID Chromosome Location
rs34112331 Chr.1: 179101780-179101780 on Build GRCh38
rs886542654 Chr.1: 179101421-179101421 on Build GRCh38
rs573591926 Chr.1: 179101437-179101437 on Build GRCh38
rs1000128412 Chr.1: 179101465-179101465 on Build GRCh38
rs377077096 Chr.1: 179101371-179101371 on Build GRCh38
rs796489798 Chr.1: 179101378-179101378 on Build GRCh38
rs71108091 Chr.1: 179101377-179101378 on Build GRCh38
rs900950865 Chr.1: 179101438-179101438 on Build GRCh38
rs891744601 Chr.1: 179101812-179101812 on Build GRCh38
rs971183740 Chr.1: 179101819-179101819 on Build GRCh38
rs76656490 Chr.1: 179101802-179101802 on Build GRCh38
rs955799193 Chr.1: 179101528-179101528 on Build GRCh38
rs939652335 Chr.1: 179101684-179101684 on Build GRCh38
rs1048500758 Chr.1: 179101431-179101431 on Build GRCh38
rs1028178134 Chr.1: 179101463-179101463 on Build GRCh38
rs926819735 Chr.1: 179101625-179101625 on Build GRCh38
rs908826755 Chr.1: 179101682-179101682 on Build GRCh38
rs749469517 Chr.1: 179101464-179101464 on Build GRCh38
rs200868662 Chr.1: 179101392-179101392 on Build GRCh38
rs1040758299 Chr.1: 179101689-179101689 on Build GRCh38
rs981318334 Chr.1: 179101575-179101575 on Build GRCh38
rs1034219043 Chr.1: 179101862-179101862 on Build GRCh38
rs541336871 Chr.1: 179101430-179101430 on Build GRCh38
rs1024944872 Chr.1: 179101818-179101818 on Build GRCh38
rs75924826 Chr.1: 179101390-179101390 on Build GRCh38
rs868132192 Chr.1: 179101520-179101520 on Build GRCh38
rs77578072 Chr.1: 179101379-179101379 on Build GRCh38
rs970803686 Chr.1: 179101532-179101532 on Build GRCh38
rs1025003763 Chr.1: 179101531-179101531 on Build GRCh38
rs774389737 Chr.1: 179101853-179101853 on Build GRCh38
rs535766020 Chr.1: 179101401-179101401 on Build GRCh38
rs1029685097 Chr.1: 179101474-179101474 on Build GRCh38
rs900901747 Chr.1: 179101698-179101698 on Build GRCh38
rs146015160 Chr.1: 179101643-179101643 on Build GRCh38
rs563631673 Chr.1: 179101816-179101816 on Build GRCh38
rs940752081 Chr.1: 179101795-179101795 on Build GRCh38
rs1049994414 Chr.1: 179101746-179101746 on Build GRCh38
rs373149601 Chr.1: 179101747-179101747 on Build GRCh38
rs61821666 Chr.1: 179101376-179101376 on Build GRCh38
rs191175379 Chr.1: 179101462-179101462 on Build GRCh38
rs939705031 Chr.1: 179101436-179101436 on Build GRCh38
rs530965708 Chr.1: 179101502-179101502 on Build GRCh38
rs988555698 Chr.1: 179101563-179101563 on Build GRCh38
rs927300434 Chr.1: 179101389-179101390 on Build GRCh38
rs1021457679 Chr.1: 179101585-179101585 on Build GRCh38
rs564406358 Chr.1: 179101723-179101723 on Build GRCh38
rs1032816901 Chr.1: 179101652-179101652 on Build GRCh38
rs886774862 Chr.1: 179101435-179101435 on Build GRCh38
rs931595341 Chr.1: 179101417-179101417 on Build GRCh38
rs1037813074 Chr.1: 179101801-179101801 on Build GRCh38
rs148008000 Chr.1: 179101400-179101400 on Build GRCh38
rs142412482 Chr.1: 179101497-179101497 on Build GRCh38

View Full Product Details