The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098519.1 | NP_001091989.1 | 7 - 8 | 1374 | 119 |
NM_152759.4 | NP_689972.3 | 7 - 8 | 1263 | 119 | |
XM_017019124.1 | XP_016874613.1 | 7 - 8 | 1382 | 119 | |
GenBank mRNA | AK124107.1 | - | 9 - 10 | 1643 | 119 |
BC029245.2 | - | 6 - 7 | 1198 | 119 |