Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC51A
Entrez Gene ID
200931
Gene Name
solute carrier family 51 alpha subunit
Gene Aliases
OSTA, OSTalpha
Chromosome Location
Chr.3: 196216512 - 196233430 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152672.5 NP_689885.4 8 - 9 1089 88
GenBank mRNA AK172837.1 - 8 - 9 1092 88
AY194243.1 - 8 - 9 910 88
BC029606.1 - 4 - 5 701 88

View Full Product Details