The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135050.1 | NP_001128522.1 | 7 - 8 | 1013 | 114 |
NM_020789.3 | NP_065840.2 | 7 - 8 | 1013 | 114 | |
XM_017001884.1 | XP_016857373.1 | 7 - 8 | 1078 | 114 | |
GenBank mRNA | AB037776.1 | - | 7 - 8 | 1013 | 114 |
AY358128.1 | - | 7 - 8 | 1031 | 114 | |
BC030141.2 | - | 7 - 8 | 1013 | 114 |