Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DIP2B
Entrez Gene ID
57609
Gene Name
disco interacting protein 2 homolog B
Gene Aliases
-
Chromosome Location
Chr.12: 50504985 - 50748667 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173602.2 NP_775873.2 24 - 25 3121 80
XM_005269044.1 XP_005269101.1 24 - 25 3124 80
XM_006719520.3 XP_006719583.1 25 - 26 3097 80
XM_006719521.3 XP_006719584.1 24 - 25 2934 80
XM_011538594.2 XP_011536896.1 25 - 26 3150 80
XM_011538595.2 XP_011536897.1 23 - 24 2982 80
XM_011538596.2 XP_011536898.1 24 - 25 2937 80
XM_017019699.1 XP_016875188.1 25 - 26 3132 80
XM_017019700.1 XP_016875189.1 24 - 25 3106 80
XM_017019701.1 XP_016875190.1 24 - 25 3103 80
XM_017019702.1 XP_016875191.1 22 - 23 2953 80
XM_017019703.1 XP_016875192.1 23 - 24 2900 80
XM_017019704.1 XP_016875193.1 23 - 24 2908 80
XM_017019705.1 XP_016875194.1 23 - 24 2890 80
GenBank mRNA AK097369.1 - 6 - 7 677 80
AK126758.1 - 13 - 14 1492 80
BC007671.2 - 8 - 9 1045 80
BC030156.2 - 8 - 9 1045 80

View Full Product Details