The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319075.1 | NP_001306004.1 | 1 - 2 | 93 | 94 |
GenBank mRNA | BC030579.2 | - | 1 - 2 | 96 | 94 |