Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WBSCR28
Entrez Gene ID
135886
Gene Name
Williams-Beuren syndrome chromosome region 28
Gene Aliases
-
Chromosome Location
Chr.7: 73860848 - 73865893 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182504.3 NP_872310.2 1 - 2 105 109
GenBank mRNA AY372053.1 - 1 - 2 110 109
BC030643.2 - 1 - 2 105 109

View Full Product Details