Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98735939-98736438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS6
Entrez Gene ID
203286
Gene Name
ankyrin repeat and sterile alpha motif domain containing 6
Gene Aliases
ANKRD14, NPHP16, PKDR1, SAMD6
UniGene
Hs.406890
Gene Chromosome Location
Chr.9: 98732009-98796542 on Build GRCh38
Transcripts
NM_173551.4 XM_005251793.4 XM_005251794.4 XM_006716998.3 XM_006716999.2 XM_011518357.2 XM_017014445.1 XM_017014446.1 XM_017014447.1

SNP Detail

SNP ID Chromosome Location
rs902745807 Chr.9: 98736335-98736335 on Build GRCh38
rs866215199 Chr.9: 98736111-98736111 on Build GRCh38
rs905084216 Chr.9: 98736399-98736399 on Build GRCh38
rs1038819569 Chr.9: 98736059-98736059 on Build GRCh38
rs1002108412 Chr.9: 98736415-98736415 on Build GRCh38
rs990225647 Chr.9: 98736234-98736234 on Build GRCh38
rs765181834 Chr.9: 98736201-98736201 on Build GRCh38
rs563957549 Chr.9: 98736126-98736126 on Build GRCh38
rs1021216208 Chr.9: 98736115-98736115 on Build GRCh38
rs560008608 Chr.9: 98736178-98736178 on Build GRCh38
rs41283622 Chr.9: 98736058-98736058 on Build GRCh38
rs1045589707 Chr.9: 98736001-98736001 on Build GRCh38
rs1024806160 Chr.9: 98736164-98736164 on Build GRCh38
rs534368552 Chr.9: 98736356-98736356 on Build GRCh38
rs184307905 Chr.9: 98736211-98736211 on Build GRCh38
rs1013242984 Chr.9: 98736163-98736163 on Build GRCh38
rs188746666 Chr.9: 98736416-98736416 on Build GRCh38
rs115752396 Chr.9: 98736057-98736057 on Build GRCh38
rs544371705 Chr.9: 98736318-98736318 on Build GRCh38
rs1046060379 Chr.9: 98736398-98736398 on Build GRCh38
rs967023925 Chr.9: 98736130-98736130 on Build GRCh38
rs779814342 Chr.9: 98736306-98736306 on Build GRCh38
rs337585 Chr.9: 98736257-98736257 on Build GRCh38
rs544269507 Chr.9: 98736113-98736113 on Build GRCh38
rs184443656 Chr.9: 98735965-98735965 on Build GRCh38
rs112795046 Chr.9: 98736174-98736174 on Build GRCh38
rs1038764873 Chr.9: 98736315-98736315 on Build GRCh38
rs1039888405 Chr.9: 98736098-98736098 on Build GRCh38
rs972747098 Chr.9: 98736215-98736215 on Build GRCh38
rs572923790 Chr.9: 98736050-98736050 on Build GRCh38
rs61383603 Chr.9: 98736327-98736327 on Build GRCh38
rs886765962 Chr.9: 98736142-98736142 on Build GRCh38
rs1040385975 Chr.9: 98736344-98736344 on Build GRCh38
rs549116363 Chr.9: 98736223-98736223 on Build GRCh38
rs946065655 Chr.9: 98736042-98736042 on Build GRCh38
rs111855758 Chr.9: 98736389-98736389 on Build GRCh38
rs567874743 Chr.9: 98736380-98736380 on Build GRCh38
rs75690065 Chr.9: 98736320-98736320 on Build GRCh38
rs949400809 Chr.9: 98736001-98736001 on Build GRCh38
rs1001430403 Chr.9: 98736157-98736157 on Build GRCh38
rs10760479 Chr.9: 98736282-98736282 on Build GRCh38
rs148654867 Chr.9: 98736112-98736112 on Build GRCh38
rs116189080 Chr.9: 98736092-98736092 on Build GRCh38
rs142157174 Chr.9: 98736167-98736167 on Build GRCh38
rs1049347736 Chr.9: 98736226-98736226 on Build GRCh38
rs7038922 Chr.9: 98736027-98736027 on Build GRCh38
rs990836554 Chr.9: 98736177-98736177 on Build GRCh38
rs572773577 Chr.9: 98736183-98736183 on Build GRCh38
rs919182586 Chr.9: 98736008-98736008 on Build GRCh38
rs1013171264 Chr.9: 98736394-98736394 on Build GRCh38
rs879263835 Chr.9: 98736162-98736162 on Build GRCh38
rs1033022427 Chr.9: 98736369-98736369 on Build GRCh38
rs945992009 Chr.9: 98736281-98736281 on Build GRCh38
rs548150946 Chr.9: 98736377-98736377 on Build GRCh38
rs151176139 Chr.9: 98736382-98736382 on Build GRCh38

View Full Product Details