The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198514.3 | NP_940916.2 | 6 - 7 | 1385 | 74 |
XM_011539769.2 | XP_011538071.1 | 6 - 7 | 1385 | 74 | |
GenBank mRNA | AK090631.1 | - | 6 - 7 | 1356 | 74 |
AK098487.1 | - | 4 - 5 | 499 | 74 | |
BC032598.1 | - | 2 - 3 | 149 | 74 | |
BX538287.1 | - | 2 - 3 | 193 | 74 | |
BX647641.1 | - | 4 - 5 | 756 | 74 |