Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMAIP1
Entrez Gene ID
5366
Gene Name
phorbol-12-myristate-13-acetate-induced protein 1
Gene Aliases
APR, NOXA
Chromosome Location
Chr.18: 59899960 - 59904306 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC032663.1 - 2 - 3 387 59
KU178263.1 - 2 - 3 214 59

View Full Product Details