Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 104782987-104783529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA1
Entrez Gene ID
19
Gene Name
ATP binding cassette subfamily A member 1
Gene Aliases
ABC-1, ABC1, CERP, HDLDT1, TGD
UniGene
Hs.659274
Gene Chromosome Location
Chr.9: 104781002-104928246 on Build GRCh38
Transcripts
NM_005502.3 XM_005251773.2 XM_005251776.2 XM_011518339.2 XM_011518340.2 XM_011518341.2 XM_011518342.2 XM_017014378.1 XM_017014379.1 XM_017014380.1 XM_017014381.1 XM_017014382.1

SNP Detail

SNP ID Chromosome Location
rs866428233 Chr.9: 104783447-104783447 on Build GRCh38
rs755419992 Chr.9: 104783462-104783462 on Build GRCh38
rs904209951 Chr.9: 104783190-104783191 on Build GRCh38
rs568125541 Chr.9: 104783113-104783113 on Build GRCh38
rs539558014 Chr.9: 104783441-104783442 on Build GRCh38
rs971912904 Chr.9: 104783427-104783427 on Build GRCh38
rs561383240 Chr.9: 104783062-104783062 on Build GRCh38
rs183535818 Chr.9: 104783474-104783474 on Build GRCh38
rs571967542 Chr.9: 104783135-104783135 on Build GRCh38
rs1037309867 Chr.9: 104783284-104783284 on Build GRCh38
rs148609447 Chr.9: 104783055-104783055 on Build GRCh38
rs1024381427 Chr.9: 104783423-104783423 on Build GRCh38
rs528656411 Chr.9: 104783097-104783097 on Build GRCh38
rs954094495 Chr.9: 104783310-104783310 on Build GRCh38
rs555000737 Chr.9: 104783492-104783492 on Build GRCh38
rs1057393470 Chr.9: 104783130-104783130 on Build GRCh38
rs1012936854 Chr.9: 104783402-104783402 on Build GRCh38
rs544667840 Chr.9: 104783137-104783137 on Build GRCh38
rs1056929833 Chr.9: 104783417-104783417 on Build GRCh38
rs547220092 Chr.9: 104783104-104783104 on Build GRCh38
rs189826506 Chr.9: 104783259-104783259 on Build GRCh38
rs962672002 Chr.9: 104783357-104783357 on Build GRCh38
rs886063300 Chr.9: 104783440-104783440 on Build GRCh38
rs1022600835 Chr.9: 104783154-104783154 on Build GRCh38
rs995391814 Chr.9: 104783332-104783332 on Build GRCh38
rs929247205 Chr.9: 104783407-104783407 on Build GRCh38
rs766398290 Chr.9: 104783189-104783189 on Build GRCh38
rs186900352 Chr.9: 104783161-104783161 on Build GRCh38
rs1031911804 Chr.9: 104783249-104783249 on Build GRCh38
rs1012677356 Chr.9: 104783153-104783153 on Build GRCh38
rs1000828107 Chr.9: 104783220-104783220 on Build GRCh38
rs753919065 Chr.9: 104783235-104783235 on Build GRCh38
rs886063299 Chr.9: 104783111-104783111 on Build GRCh38
rs958673244 Chr.9: 104783399-104783399 on Build GRCh38
rs1037635917 Chr.9: 104783093-104783093 on Build GRCh38
rs565619592 Chr.9: 104783383-104783383 on Build GRCh38
rs569604699 Chr.9: 104783441-104783441 on Build GRCh38
rs1033601541 Chr.9: 104783360-104783360 on Build GRCh38
rs898402597 Chr.9: 104783311-104783311 on Build GRCh38
rs547038401 Chr.9: 104783024-104783024 on Build GRCh38
rs998362887 Chr.9: 104783269-104783269 on Build GRCh38
rs977615117 Chr.9: 104783428-104783428 on Build GRCh38

View Full Product Details