Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 104783844-104784348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA1
Entrez Gene ID
19
Gene Name
ATP binding cassette subfamily A member 1
Gene Aliases
ABC-1, ABC1, CERP, HDLDT1, TGD
UniGene
Hs.659274
Gene Chromosome Location
Chr.9: 104781002-104928246 on Build GRCh38
Transcripts
NM_005502.3 XM_005251773.2 XM_005251776.2 XM_011518339.2 XM_011518340.2 XM_011518341.2 XM_011518342.2 XM_017014378.1 XM_017014379.1 XM_017014380.1 XM_017014381.1 XM_017014382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1459098 c.6779A>G p.Y2260C Chr.9: 104784322-104784322 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779593454 Chr.9: 104784298-104784298 on Build GRCh38
rs754345926 Chr.9: 104783967-104783967 on Build GRCh38
rs1024211015 Chr.9: 104783935-104783935 on Build GRCh38
rs774328251 Chr.9: 104784028-104784028 on Build GRCh38
rs969929677 Chr.9: 104784248-104784248 on Build GRCh38
rs1045523810 Chr.9: 104784011-104784011 on Build GRCh38
rs561319670 Chr.9: 104783898-104783898 on Build GRCh38
rs764486276 Chr.9: 104784273-104784273 on Build GRCh38
rs200582190 Chr.9: 104783993-104783993 on Build GRCh38
rs538474704 Chr.9: 104784240-104784240 on Build GRCh38
rs557492263 Chr.9: 104783994-104783994 on Build GRCh38
rs754415338 Chr.9: 104784266-104784267 on Build GRCh38
rs751049570 Chr.9: 104784318-104784318 on Build GRCh38
rs748823432 Chr.9: 104783962-104783962 on Build GRCh38
rs1050756559 Chr.9: 104784071-104784071 on Build GRCh38
rs886063303 Chr.9: 104784026-104784026 on Build GRCh38
rs1046318975 Chr.9: 104784048-104784048 on Build GRCh38
rs34352903 Chr.9: 104784116-104784117 on Build GRCh38
rs1011696174 Chr.9: 104784158-104784158 on Build GRCh38
rs375241409 Chr.9: 104784295-104784295 on Build GRCh38
rs73517867 Chr.9: 104783899-104783899 on Build GRCh38
rs867969689 Chr.9: 104783936-104783936 on Build GRCh38
rs777237083 Chr.9: 104784275-104784275 on Build GRCh38
rs779300353 Chr.9: 104784306-104784306 on Build GRCh38
rs557180532 Chr.9: 104784283-104784283 on Build GRCh38
rs762785983 Chr.9: 104784299-104784299 on Build GRCh38
rs886063304 Chr.9: 104784183-104784183 on Build GRCh38
rs895062056 Chr.9: 104784026-104784026 on Build GRCh38
rs147610260 Chr.9: 104784079-104784079 on Build GRCh38
rs907173855 Chr.9: 104784063-104784063 on Build GRCh38
rs73517870 Chr.9: 104783920-104783920 on Build GRCh38
rs988979584 Chr.9: 104784007-104784007 on Build GRCh38
rs1026138617 Chr.9: 104784163-104784163 on Build GRCh38
rs1008859316 Chr.9: 104784265-104784265 on Build GRCh38
rs764422600 Chr.9: 104783950-104783950 on Build GRCh38
rs13306080 Chr.9: 104783947-104783947 on Build GRCh38
rs763013834 Chr.9: 104783993-104783994 on Build GRCh38
rs780958155 Chr.9: 104784321-104784321 on Build GRCh38
rs1032159491 Chr.9: 104784082-104784082 on Build GRCh38
rs759527890 Chr.9: 104784030-104784030 on Build GRCh38
rs1038459626 Chr.9: 104784137-104784137 on Build GRCh38
rs151170184 Chr.9: 104783909-104783909 on Build GRCh38
rs913010213 Chr.9: 104784017-104784017 on Build GRCh38
rs79318417 Chr.9: 104783995-104783995 on Build GRCh38
rs370983511 Chr.9: 104784293-104784293 on Build GRCh38
rs542665134 Chr.9: 104783867-104783867 on Build GRCh38
rs552730637 Chr.9: 104784182-104784182 on Build GRCh38
rs1017089456 Chr.9: 104783866-104783866 on Build GRCh38
rs894316609 Chr.9: 104783983-104783983 on Build GRCh38
rs77096626 Chr.9: 104783996-104783996 on Build GRCh38
rs575454176 Chr.9: 104784289-104784289 on Build GRCh38
rs898676349 Chr.9: 104784150-104784150 on Build GRCh38
rs146987516 Chr.9: 104783973-104783973 on Build GRCh38
rs933632092 Chr.9: 104784032-104784033 on Build GRCh38
rs972456579 Chr.9: 104784304-104784304 on Build GRCh38
rs548612176 Chr.9: 104784052-104784052 on Build GRCh38
rs961308213 Chr.9: 104784302-104784302 on Build GRCh38
rs768343668 Chr.9: 104784108-104784108 on Build GRCh38
rs925787574 Chr.9: 104783987-104783987 on Build GRCh38
rs878937680 Chr.9: 104784245-104784245 on Build GRCh38
rs530474404 Chr.9: 104784012-104784012 on Build GRCh38
rs569460168 Chr.9: 104783978-104783978 on Build GRCh38
rs372148712 Chr.9: 104784299-104784299 on Build GRCh38
rs755818735 Chr.9: 104783917-104783917 on Build GRCh38
rs74316246 Chr.9: 104784219-104784219 on Build GRCh38
rs367822841 Chr.9: 104784178-104784178 on Build GRCh38
rs370948252 Chr.9: 104783932-104783932 on Build GRCh38
rs368288959 Chr.9: 104784296-104784296 on Build GRCh38
rs748194153 Chr.9: 104783993-104783993 on Build GRCh38

View Full Product Details