The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152911.3 | NP_690875.1 | 3 - 4 | 948 | 63 |
NM_207128.2 | NP_997011.1 | 3 - 4 | 948 | 63 | |
GenBank mRNA | AF226657.1 | - | 2 - 3 | 688 | 63 |
AF312698.1 | - | 3 - 4 | 994 | 63 | |
AK128001.1 | - | 3 - 4 | 832 | 63 | |
AL834535.1 | - | 1 - 2 | 193 | 63 | |
AY358418.1 | - | 3 - 4 | 888 | 63 | |
AY541516.1 | - | 3 - 4 | 924 | 63 | |
AY541520.1 | - | - | 1312 | 63 | |
BC032778.1 | - | 3 - 4 | 959 | 63 |