The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001097638.2 | NP_001091107.1 | 1 - 2 | 110 | 62 |
GenBank mRNA | AK290277.1 | - | 1 - 2 | 106 | 62 |
BC001899.1 | - | 1 - 2 | 60 | 62 | |
BC121066.1 | - | 1 - 2 | 42 | 62 | |
BE266792.1 | - | 1 - 2 | 42 | 62 | |
DA918525.1 | - | 1 - 2 | 110 | 62 |