The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300980.1 | NP_001287909.1 | 7 - 8 | 928 | 61 |
NM_001300982.1 | NP_001287911.1 | 5 - 6 | 617 | 61 | |
NM_018502.4 | NP_060972.3 | 7 - 8 | 910 | 61 | |
XM_005268477.1 | XP_005268534.1 | 7 - 8 | 913 | 61 | |
XM_011537663.2 | XP_011535965.1 | 7 - 8 | 928 | 61 | |
XM_011537665.2 | XP_011535967.1 | 6 - 7 | 992 | 61 | |
XM_011537668.2 | XP_011535970.1 | 5 - 6 | 665 | 61 | |
XM_017009617.1 | XP_016865106.1 | 7 - 8 | 910 | 61 | |
XM_017009618.1 | XP_016865107.1 | 7 - 8 | 928 | 61 | |
XM_017009619.1 | XP_016865108.1 | 6 - 7 | 716 | 61 | |
XM_017009620.1 | XP_016865109.1 | 6 - 7 | 701 | 61 | |
GenBank mRNA | AK097088.1 | - | 7 - 8 | 874 | 61 |
AK289495.1 | - | 5 - 6 | 575 | 61 | |
BC001910.1 | - | 7 - 8 | 851 | 61 | |
BC009618.1 | - | 2 - 3 | 143 | 61 |