The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282546.1 | NP_001269475.1 | 7 - 8 | 968 | 76 |
NM_001282547.1 | NP_001269476.1 | 7 - 8 | 953 | 76 | |
NM_032017.2 | NP_114406.1 | 8 - 9 | 1190 | 76 | |
GenBank mRNA | AK024504.1 | - | 6 - 7 | 752 | 76 |
AK075029.1 | - | 4 - 5 | 469 | 76 | |
AK075048.1 | - | 7 - 8 | 947 | 76 | |
AL834137.1 | - | 8 - 9 | 1147 | 76 | |
AY198395.1 | - | 7 - 8 | 923 | 76 | |
BC005169.2 | - | 2 - 3 | 152 | 76 | |
BC007835.2 | - | 7 - 8 | 921 | 76 | |
BC008344.1 | - | 7 - 8 | 929 | 76 |