Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NPPA
- Entrez Gene ID
-
4878
- Gene Name
- natriuretic peptide A
- Gene Aliases
-
ANF, ANP, ATFB6, ATRST2, CDD, CDD-ANF, CDP, PND
- Chromosome Location
-
Chr.1: 11845709 - 11847783 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC005893.1 |
- |
2 - 3 |
550 |
55 |
View Full Product Details