Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 72925370-72925874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH1A1
Entrez Gene ID
216
Gene Name
aldehyde dehydrogenase 1 family member A1
Gene Aliases
ALDC, ALDH-E1, ALDH1, ALDH11, HEL-9, HEL-S-53e, HEL12, PUMB1, RALDH1
UniGene
Hs.76392
Gene Chromosome Location
Chr.9: 72900662-72953317 on Build GRCh38
Transcripts
NM_000689.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6210621 c.655G>A p.V219M Chr.9: 72925504-72925504 on Build GRCh38
4552488 c.602G>A p.G201E Chr.9: 72925557-72925557 on Build GRCh38
6517148 c.520G>T p.V174F Chr.9: 72925597-72925597 on Build GRCh38
456064 c.579G>A p.K193K Chr.9: 72925580-72925580 on Build GRCh38
6456502 c.617C>T p.A206V Chr.9: 72925500-72925500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760357552 Chr.9: 72925499-72925499 on Build GRCh38
rs1012148168 Chr.9: 72925798-72925798 on Build GRCh38
rs1023460281 Chr.9: 72925493-72925493 on Build GRCh38
rs746834602 Chr.9: 72925573-72925573 on Build GRCh38
rs975431872 Chr.9: 72925694-72925694 on Build GRCh38
rs2288086 Chr.9: 72925656-72925656 on Build GRCh38
rs1049150881 Chr.9: 72925423-72925423 on Build GRCh38
rs761319210 Chr.9: 72925473-72925473 on Build GRCh38
rs763683870 Chr.9: 72925505-72925505 on Build GRCh38
rs183999437 Chr.9: 72925782-72925782 on Build GRCh38
rs774798542 Chr.9: 72925650-72925650 on Build GRCh38
rs561859585 Chr.9: 72925810-72925810 on Build GRCh38
rs781178553 Chr.9: 72925811-72925811 on Build GRCh38
rs555469171 Chr.9: 72925560-72925560 on Build GRCh38
rs566343527 Chr.9: 72925427-72925427 on Build GRCh38
rs146885014 Chr.9: 72925568-72925568 on Build GRCh38
rs376533427 Chr.9: 72925472-72925472 on Build GRCh38
rs372664748 Chr.9: 72925587-72925587 on Build GRCh38
rs560735521 Chr.9: 72925717-72925717 on Build GRCh38
rs771883684 Chr.9: 72925466-72925466 on Build GRCh38
rs759915629 Chr.9: 72925653-72925653 on Build GRCh38
rs535525647 Chr.9: 72925689-72925689 on Build GRCh38
rs8187930 Chr.9: 72925477-72925477 on Build GRCh38
rs751432901 Chr.9: 72925736-72925736 on Build GRCh38
rs776875387 Chr.9: 72925467-72925467 on Build GRCh38
rs944840027 Chr.9: 72925694-72925694 on Build GRCh38
rs750444421 Chr.9: 72925554-72925554 on Build GRCh38
rs940810006 Chr.9: 72925774-72925774 on Build GRCh38
rs375335307 Chr.9: 72925690-72925690 on Build GRCh38
rs544635792 Chr.9: 72925648-72925648 on Build GRCh38
rs896507121 Chr.9: 72925787-72925787 on Build GRCh38
rs367856349 Chr.9: 72925752-72925752 on Build GRCh38
rs540951235 Chr.9: 72925716-72925716 on Build GRCh38
rs773325316 Chr.9: 72925601-72925601 on Build GRCh38
rs748201834 Chr.9: 72925470-72925470 on Build GRCh38
rs539350021 Chr.9: 72925417-72925417 on Build GRCh38
rs8187931 Chr.9: 72925420-72925420 on Build GRCh38
rs878873028 Chr.9: 72925480-72925480 on Build GRCh38
rs879141810 Chr.9: 72925478-72925479 on Build GRCh38
rs192915962 Chr.9: 72925750-72925750 on Build GRCh38
rs904955542 Chr.9: 72925832-72925832 on Build GRCh38
rs868244622 Chr.9: 72925421-72925421 on Build GRCh38
rs577923942 Chr.9: 72925711-72925711 on Build GRCh38
rs753473268 Chr.9: 72925512-72925512 on Build GRCh38
rs530527162 Chr.9: 72925830-72925830 on Build GRCh38
rs747288253 Chr.9: 72925451-72925451 on Build GRCh38
rs142547395 Chr.9: 72925548-72925548 on Build GRCh38
rs1000594311 Chr.9: 72925555-72925555 on Build GRCh38
rs907966170 Chr.9: 72925757-72925757 on Build GRCh38
rs769827616 Chr.9: 72925597-72925597 on Build GRCh38
rs1004892060 Chr.9: 72925435-72925435 on Build GRCh38
rs774675922 Chr.9: 72925491-72925491 on Build GRCh38
rs766318889 Chr.9: 72925723-72925723 on Build GRCh38
rs8187929 Chr.9: 72925588-72925588 on Build GRCh38
rs200452607 Chr.9: 72925577-72925577 on Build GRCh38
rs770966677 Chr.9: 72925567-72925567 on Build GRCh38
rs1003764136 Chr.9: 72925841-72925841 on Build GRCh38
rs764111712 Chr.9: 72925796-72925796 on Build GRCh38
rs372689208 Chr.9: 72925471-72925471 on Build GRCh38
rs755077130 Chr.9: 72925801-72925801 on Build GRCh38
rs147729972 Chr.9: 72925547-72925547 on Build GRCh38
rs756725946 Chr.9: 72925524-72925524 on Build GRCh38
rs779982428 Chr.9: 72925439-72925439 on Build GRCh38
rs768212750 Chr.9: 72925402-72925402 on Build GRCh38
rs768972884 Chr.9: 72925463-72925463 on Build GRCh38
rs866644556 Chr.9: 72925501-72925501 on Build GRCh38
rs767857748 Chr.9: 72925655-72925655 on Build GRCh38
rs910641935 Chr.9: 72925678-72925678 on Build GRCh38
rs760776363 Chr.9: 72925767-72925767 on Build GRCh38
rs879192228 Chr.9: 72925482-72925482 on Build GRCh38
rs749842101 Chr.9: 72925640-72925640 on Build GRCh38

View Full Product Details