Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 87497576-87498070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAPK1
Entrez Gene ID
1612
Gene Name
death associated protein kinase 1
Gene Aliases
DAPK
UniGene
Hs.380277
Gene Chromosome Location
Chr.9: 87497228-87708634 on Build GRCh38
Transcripts
NM_001288730.1 NM_001288731.1 NM_004938.3 XM_005251757.3

SNP Detail

SNP ID Chromosome Location
rs17515986 Chr.9: 87498026-87498026 on Build GRCh38
rs543349629 Chr.9: 87498040-87498040 on Build GRCh38
rs985214031 Chr.9: 87497782-87497782 on Build GRCh38
rs951847409 Chr.9: 87497799-87497799 on Build GRCh38
rs939014835 Chr.9: 87497712-87497712 on Build GRCh38
rs1043236766 Chr.9: 87497721-87497721 on Build GRCh38
rs960071635 Chr.9: 87497664-87497664 on Build GRCh38
rs1048282066 Chr.9: 87498039-87498039 on Build GRCh38
rs542742912 Chr.9: 87497640-87497640 on Build GRCh38
rs1049128614 Chr.9: 87498051-87498051 on Build GRCh38
rs905959067 Chr.9: 87497646-87497646 on Build GRCh38
rs913502150 Chr.9: 87497707-87497707 on Build GRCh38
rs1006743514 Chr.9: 87497910-87497910 on Build GRCh38
rs965017089 Chr.9: 87497800-87497800 on Build GRCh38
rs778982179 Chr.9: 87497631-87497631 on Build GRCh38
rs926308474 Chr.9: 87497709-87497709 on Build GRCh38
rs559842437 Chr.9: 87497653-87497653 on Build GRCh38
rs913451211 Chr.9: 87497822-87497822 on Build GRCh38
rs1011768526 Chr.9: 87497606-87497606 on Build GRCh38
rs1004811683 Chr.9: 87497876-87497876 on Build GRCh38
rs370041930 Chr.9: 87497874-87497874 on Build GRCh38
rs1015494900 Chr.9: 87497670-87497670 on Build GRCh38
rs919642252 Chr.9: 87497684-87497684 on Build GRCh38
rs904690568 Chr.9: 87497731-87497731 on Build GRCh38
rs972140745 Chr.9: 87497789-87497789 on Build GRCh38
rs960694071 Chr.9: 87497906-87497906 on Build GRCh38
rs2302791 Chr.9: 87497978-87497978 on Build GRCh38
rs545388283 Chr.9: 87497706-87497706 on Build GRCh38
rs1056628443 Chr.9: 87497719-87497719 on Build GRCh38
rs911957724 Chr.9: 87497655-87497655 on Build GRCh38
rs965130584 Chr.9: 87497928-87497928 on Build GRCh38
rs987982605 Chr.9: 87497648-87497648 on Build GRCh38
rs1049188707 Chr.9: 87497957-87497957 on Build GRCh38
rs762645135 Chr.9: 87497941-87497941 on Build GRCh38
rs565303904 Chr.9: 87497740-87497740 on Build GRCh38
rs995428196 Chr.9: 87497904-87497904 on Build GRCh38
rs774403833 Chr.9: 87497980-87497980 on Build GRCh38
rs936181246 Chr.9: 87497911-87497911 on Build GRCh38
rs954047136 Chr.9: 87497778-87497778 on Build GRCh38
rs1053343695 Chr.9: 87497701-87497701 on Build GRCh38
rs528598723 Chr.9: 87497675-87497675 on Build GRCh38
rs1036329253 Chr.9: 87497892-87497892 on Build GRCh38
rs979033151 Chr.9: 87497992-87497992 on Build GRCh38
rs1018986998 Chr.9: 87497882-87497882 on Build GRCh38
rs887820450 Chr.9: 87497962-87497962 on Build GRCh38
rs988487257 Chr.9: 87497818-87497818 on Build GRCh38
rs992970719 Chr.9: 87497981-87497981 on Build GRCh38
rs930588676 Chr.9: 87497697-87497697 on Build GRCh38
rs533245312 Chr.9: 87497742-87497742 on Build GRCh38
rs897814110 Chr.9: 87497885-87497885 on Build GRCh38
rs932420230 Chr.9: 87497688-87497688 on Build GRCh38
rs1001626107 Chr.9: 87497736-87497736 on Build GRCh38
rs925036620 Chr.9: 87497993-87497993 on Build GRCh38
rs1050578106 Chr.9: 87497921-87497921 on Build GRCh38
rs980565119 Chr.9: 87497897-87497897 on Build GRCh38
rs575752217 Chr.9: 87498036-87498036 on Build GRCh38
rs574146747 Chr.9: 87497991-87497991 on Build GRCh38
rs1026816500 Chr.9: 87497775-87497775 on Build GRCh38
rs1000432020 Chr.9: 87497733-87497733 on Build GRCh38
rs957336788 Chr.9: 87497809-87497837 on Build GRCh38
rs377011195 Chr.9: 87497994-87497994 on Build GRCh38
rs985652769 Chr.9: 87497690-87497690 on Build GRCh38
rs943713953 Chr.9: 87497700-87497700 on Build GRCh38
rs968825113 Chr.9: 87497939-87497939 on Build GRCh38
rs573347820 Chr.9: 87498001-87498001 on Build GRCh38
rs550136479 Chr.9: 87497743-87497743 on Build GRCh38
rs1050650212 Chr.9: 87497871-87497871 on Build GRCh38
rs1035658671 Chr.9: 87497656-87497656 on Build GRCh38
rs1015442753 Chr.9: 87497770-87497770 on Build GRCh38
rs1004864001 Chr.9: 87497758-87497758 on Build GRCh38
rs899199670 Chr.9: 87497903-87497903 on Build GRCh38
rs973952268 Chr.9: 87497683-87497683 on Build GRCh38
rs541591442 Chr.9: 87497920-87497920 on Build GRCh38
rs947511521 Chr.9: 87497720-87497720 on Build GRCh38
rs1043441721 Chr.9: 87497850-87497850 on Build GRCh38
rs937537575 Chr.9: 87497858-87497858 on Build GRCh38
rs1043744230 Chr.9: 87497845-87497845 on Build GRCh38
rs1022102661 Chr.9: 87497630-87497630 on Build GRCh38
rs1019343475 Chr.9: 87497915-87497915 on Build GRCh38
rs565730073 Chr.9: 87497907-87497907 on Build GRCh38
rs1009317477 Chr.9: 87497877-87497877 on Build GRCh38
rs775286116 Chr.9: 87497797-87497797 on Build GRCh38
rs992031742 Chr.9: 87497667-87497667 on Build GRCh38
rs553297705 Chr.9: 87497999-87497999 on Build GRCh38
rs905906265 Chr.9: 87497725-87497725 on Build GRCh38
rs534426669 Chr.9: 87497909-87497909 on Build GRCh38
rs988434941 Chr.9: 87497930-87497930 on Build GRCh38
rs1013526573 Chr.9: 87497987-87497987 on Build GRCh38
rs904723551 Chr.9: 87497859-87497859 on Build GRCh38
rs912224093 Chr.9: 87497691-87497691 on Build GRCh38
rs912895921 Chr.9: 87497933-87497933 on Build GRCh38
rs1008871639 Chr.9: 87497768-87497768 on Build GRCh38
rs979026269 Chr.9: 87497955-87497955 on Build GRCh38
rs1057125822 Chr.9: 87497863-87497863 on Build GRCh38
rs36228007 Chr.9: 87497965-87497965 on Build GRCh38
rs529133517 Chr.9: 87497825-87497825 on Build GRCh38
rs961333508 Chr.9: 87497771-87497771 on Build GRCh38
rs945011911 Chr.9: 87497935-87497935 on Build GRCh38
rs961177442 Chr.9: 87497681-87497681 on Build GRCh38
rs927478611 Chr.9: 87497848-87497848 on Build GRCh38
rs953391250 Chr.9: 87497613-87497613 on Build GRCh38
rs1027725291 Chr.9: 87497893-87497893 on Build GRCh38
rs1041906126 Chr.9: 87497728-87497735 on Build GRCh38
rs569741548 Chr.9: 87497750-87497750 on Build GRCh38
rs1041016000 Chr.9: 87497936-87497936 on Build GRCh38
rs1029336730 Chr.9: 87497665-87497675 on Build GRCh38
rs533943949 Chr.9: 87497816-87497822 on Build GRCh38
rs557672833 Chr.9: 87497990-87497990 on Build GRCh38
rs995707360 Chr.9: 87497773-87497773 on Build GRCh38
rs1033551030 Chr.9: 87497972-87497972 on Build GRCh38

View Full Product Details