Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 87641862-87642403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAPK1
Entrez Gene ID
1612
Gene Name
death associated protein kinase 1
Gene Aliases
DAPK
UniGene
Hs.380277
Gene Chromosome Location
Chr.9: 87497228-87708634 on Build GRCh38
Transcripts
NM_001288729.1 NM_001288730.1 NM_001288731.1 NM_004938.3 XM_005251757.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
456197 c.866C>G p.S289* Chr.9: 87642006-87642006 on Build GRCh38
4615058 c.906_907insA p.W305fs*23 Chr.9: 87642046-87642047 on Build GRCh38
7205727 c.913_914insA p.W305fs*1 Chr.9: 87642053-87642054 on Build GRCh38
4794864 c.902C>T p.A301V Chr.9: 87642042-87642042 on Build GRCh38
243956 c.894G>A p.K298K Chr.9: 87642034-87642034 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969722015 Chr.9: 87641917-87641917 on Build GRCh38
rs764735226 Chr.9: 87642098-87642098 on Build GRCh38
rs891034396 Chr.9: 87642151-87642151 on Build GRCh38
rs36212049 Chr.9: 87642133-87642133 on Build GRCh38
rs983613479 Chr.9: 87642028-87642028 on Build GRCh38
rs1008013251 Chr.9: 87641898-87641898 on Build GRCh38
rs776082970 Chr.9: 87642069-87642069 on Build GRCh38
rs370702960 Chr.9: 87642280-87642280 on Build GRCh38
rs868463663 Chr.9: 87642038-87642038 on Build GRCh38
rs373351220 Chr.9: 87641948-87641948 on Build GRCh38
rs773441662 Chr.9: 87642164-87642164 on Build GRCh38
rs529654079 Chr.9: 87642130-87642130 on Build GRCh38
rs36212048 Chr.9: 87641914-87641914 on Build GRCh38
rs368752543 Chr.9: 87641908-87641908 on Build GRCh38
rs758953837 Chr.9: 87641945-87641945 on Build GRCh38
rs768179901 Chr.9: 87642063-87642063 on Build GRCh38
rs954217491 Chr.9: 87642336-87642336 on Build GRCh38
rs769677495 Chr.9: 87641927-87641928 on Build GRCh38
rs772116415 Chr.9: 87641990-87641990 on Build GRCh38
rs750683161 Chr.9: 87641933-87641933 on Build GRCh38
rs372530198 Chr.9: 87642094-87642094 on Build GRCh38
rs922036722 Chr.9: 87642132-87642132 on Build GRCh38
rs1051754024 Chr.9: 87642145-87642145 on Build GRCh38
rs959703819 Chr.9: 87642332-87642332 on Build GRCh38
rs376381937 Chr.9: 87642154-87642154 on Build GRCh38
rs928855770 Chr.9: 87641950-87641950 on Build GRCh38
rs567285437 Chr.9: 87642340-87642340 on Build GRCh38
rs756338275 Chr.9: 87641975-87641975 on Build GRCh38
rs541858304 Chr.9: 87641906-87641906 on Build GRCh38
rs151043930 Chr.9: 87642241-87642241 on Build GRCh38
rs759051051 Chr.9: 87641923-87641924 on Build GRCh38
rs528655981 Chr.9: 87642185-87642185 on Build GRCh38
rs1001208287 Chr.9: 87642301-87642301 on Build GRCh38
rs938957032 Chr.9: 87642004-87642004 on Build GRCh38
rs891065434 Chr.9: 87641891-87641891 on Build GRCh38
rs377546052 Chr.9: 87641986-87641986 on Build GRCh38
rs866879950 Chr.9: 87642380-87642380 on Build GRCh38
rs3118846 Chr.9: 87641912-87641912 on Build GRCh38
rs765616691 Chr.9: 87641929-87641929 on Build GRCh38
rs983666289 Chr.9: 87642333-87642333 on Build GRCh38
rs369204279 Chr.9: 87641919-87641919 on Build GRCh38
rs555216707 Chr.9: 87642365-87642365 on Build GRCh38
rs866637667 Chr.9: 87641951-87641951 on Build GRCh38
rs535366720 Chr.9: 87642342-87642342 on Build GRCh38
rs902647424 Chr.9: 87642264-87642264 on Build GRCh38
rs891604358 Chr.9: 87642239-87642239 on Build GRCh38
rs551668443 Chr.9: 87642191-87642191 on Build GRCh38
rs1013798329 Chr.9: 87642240-87642240 on Build GRCh38
rs750006626 Chr.9: 87641892-87641892 on Build GRCh38
rs200137928 Chr.9: 87642034-87642034 on Build GRCh38
rs775945094 Chr.9: 87641920-87641921 on Build GRCh38
rs188260637 Chr.9: 87641955-87641955 on Build GRCh38
rs559991338 Chr.9: 87642143-87642143 on Build GRCh38
rs755075838 Chr.9: 87641956-87641956 on Build GRCh38
rs775262422 Chr.9: 87642029-87642029 on Build GRCh38
rs753081481 Chr.9: 87642071-87642071 on Build GRCh38
rs778782592 Chr.9: 87641984-87641984 on Build GRCh38
rs748404270 Chr.9: 87641974-87641974 on Build GRCh38
rs36212050 Chr.9: 87642259-87642259 on Build GRCh38
rs1035392483 Chr.9: 87642321-87642321 on Build GRCh38
rs563266740 Chr.9: 87641961-87641961 on Build GRCh38

View Full Product Details